siRNA Details
| Gene Symbol: March5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106372.2 | NP_001099842.1 | 3 | 602 | ||
| XM_008760367.1 | XP_008758589.1 | 3 | 608 | |||
| GenBank mRNA | BC158801.1 | - | 3 | 602 | 7321043 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

March5| Gene Symbol: March5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106372.2 | NP_001099842.1 | 3 | 602 | ||
| XM_008760367.1 | XP_008758589.1 | 3 | 608 | |||
| GenBank mRNA | BC158801.1 | - | 3 | 602 | 7321043 | |