siRNA Details
Gene Symbol: Ifit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024753.1 | NP_001019924.1 | 2 | 879 | ||
GenBank mRNA | BC097384.1 | - | 2 | 879 | 7446350 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ifit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024753.1 | NP_001019924.1 | 2 | 879 | ||
GenBank mRNA | BC097384.1 | - | 2 | 879 | 7446350 |