siRNA Details
| Gene Symbol: Olr385 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760585.1 | XP_008758807.1 | 2 | 761 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr385| Gene Symbol: Olr385 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760585.1 | XP_008758807.1 | 2 | 761 | ||