siRNA Details
| Gene Symbol: Taar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008512.1 | NP_001008512.1 | 2 | 956 | ||
| GenBank mRNA | AY702316.1 | - | 2 | 956 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Taar2| Gene Symbol: Taar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008512.1 | NP_001008512.1 | 2 | 956 | ||
| GenBank mRNA | AY702316.1 | - | 2 | 956 | ||
Set Membership: |
|