siRNA Details
| Gene Symbol: Taar5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009650.1 | NP_001009650.1 | 1 | 370 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001009574, BC139188, BC139191.
Set Membership: |
|

Taar5
Genomic Map