siRNA Details
| Gene Symbol: VSNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003385.4 | NP_003376.2 | 4 | 840 | ||
| GenBank mRNA | AB001104.1 | - | 4 | 595 | ||
| AF039555.1 | - | 4 | 648 | |||
| BC022012.2 | - | 4 | 677 | 4823954 | ||
| U14747.1 | - | 4 | 663 | |||

VSNL1
Genomic Map