siRNA Details
| Gene Symbol: Olr1683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000269.1 | NP_001000269.1 | 1 | 422 | ||
| Gene Symbol: Olr1670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000895.1 | NP_001000895.1 | 1 | 422 | ||
| Gene Symbol: Olr1682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000890.1 | NP_001000890.1 | 1 | 422 | ||

Olr1683
Genomic Map