siRNA Details
| Gene Symbol: LOC294154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039607.1 | NP_001034696.1 | 4 | 596 | ||
| XM_008772799.1 | XP_008771021.1 | 4 | 790 | |||
| XM_008772800.1 | XP_008771022.1 | 4 | 904 | |||
| XM_008772801.1 | XP_008771023.1 | 4 | 1329 | |||
| GenBank mRNA | BC112320.1 | - | 4 | 596 | 7933524 | |

LOC294154
Genomic Map