siRNA Details
| Gene Symbol: Olr1696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206850.1 | NP_996732.1 | 1 | 848 | ||
| GenBank mRNA | AF034896.1 | - | 1070 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1696| Gene Symbol: Olr1696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206850.1 | NP_996732.1 | 1 | 848 | ||
| GenBank mRNA | AF034896.1 | - | 1070 | |||