siRNA Details
| Gene Symbol: Olr1696 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206850.1 | NP_996732.1 | 1 | 58 | ||
| Gene Symbol: Olr1697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001111.1 | NP_001001111.1 | 1 | 58 | ||

Olr1696
Genomic Map