siRNA Details
Gene Symbol: Olr1696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206850.1 | NP_996732.1 | 1 | 654 | ||
GenBank mRNA | AF034896.1 | - | 876 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206850.1 | NP_996732.1 | 1 | 654 | ||
GenBank mRNA | AF034896.1 | - | 876 |