siRNA Details
Gene Symbol: Olr1691 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000273.1 | NP_001000273.1 | 1 | 667 |
Gene Symbol: LOC100910382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751905.2 | XP_003751953.1 | 1 | 680 |
Gene Symbol: LOC100910420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751906.2 | XP_003751954.1 | 684 |
Gene Symbol: LOC100910696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772684.1 | XP_008770906.1 | 690 |
Gene Symbol: Olr1688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000275.1 | NP_001000275.1 | 667 |
Gene Symbol: Olr1689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000889.1 | NP_001000889.1 | 1 | 667 |
Gene Symbol: Olr1690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000274.1 | NP_001000274.1 | 1 | 667 |
Gene Symbol: Olr1692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000888.1 | NP_001000888.1 | 1 | 664 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
