siRNA Details
| Gene Symbol: Olr1876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_574713.4 | XP_574713.4 | 2 | 97 | ||
| Gene Symbol: Olr1729 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255881.1 | XP_006255943.1 | 1 | 61 | ||

Olr1876
Genomic Map