siRNA Details
| Gene Symbol: Olr1731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_227934.5 | XP_227934.5 | 2 | 64 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1731| Gene Symbol: Olr1731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_227934.5 | XP_227934.5 | 2 | 64 | ||