siRNA Details
| Gene Symbol: Olr1743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001425.1 | NP_001001425.1 | 1 | 224 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1743| Gene Symbol: Olr1743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001425.1 | NP_001001425.1 | 1 | 224 | ||
Set Membership: |
|