siRNA Details
| Gene Symbol: Trim10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009176.2 | NP_001009176.1 | 6 | 1270 | ||
| GenBank mRNA | BC129078.1 | - | 6 | 1270 | 8371876 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim10| Gene Symbol: Trim10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009176.2 | NP_001009176.1 | 6 | 1270 | ||
| GenBank mRNA | BC129078.1 | - | 6 | 1270 | 8371876 | |