siRNA Details
| Gene Symbol: Gtf2h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212501.2 | NP_997666.1 | 11, 12 | 1268 | ||
| XM_006255920.2 | XP_006255982.1 | 11, 12 | 1317 | |||
| GenBank mRNA | BC083703.1 | - | 9, 10 | 1494 | ||
| BC127469.1 | - | 11, 12 | 1240 | 7324889 | ||

Gtf2h4
Genomic Map