siRNA Details
| Gene Symbol: Gtf2h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212501.2 | NP_997666.1 | 2 | 213 | ||
| XM_006255920.2 | XP_006255982.1 | 2 | 262 | |||
| GenBank mRNA | BC083703.1 | - | 2 | 171 | ||
| BC127469.1 | - | 2 | 185 | 7324889 | ||
| CK472761.1 | - | 2 | 213 | |||

Gtf2h4
Genomic Map