siRNA Details
| Gene Symbol: Ly6g6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001969.1 | NP_001001969.1 | 2 | 151 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly6g6c| Gene Symbol: Ly6g6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001969.1 | NP_001001969.1 | 2 | 151 | ||