siRNA Details
| Gene Symbol: Msh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212536.2 | NP_997701.2 | 5, 6 | 442 | ||
| XM_006256057.2 | XP_006256119.1 | 5, 6 | 1269 | |||
| XM_008772717.1 | XP_008770939.1 | 4, 5 | 1510 | |||
| XM_008772718.1 | XP_008770940.1 | 4, 5 | 1510 | |||
| GenBank mRNA | BC083904.1 | - | 5, 6 | 442 | 7133915 | |

Msh5
Genomic Map