siRNA Details
| Gene Symbol: Nelfe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165901.1 | NP_001159373.1 | 4, 5 | 366 | ||
| NM_212548.2 | NP_997713.1 | 4, 5 | 398 | |||
| XM_006255921.2 | XP_006255983.1 | 3, 4 | 527 | |||
| XM_006255922.2 | XP_006255984.1 | 4, 5 | 576 | |||
| GenBank mRNA | BC079427.1 | - | 4, 5 | 398 | 7114415 | |
| CO401737.1 | - | 4, 5 | 366 | |||

Nelfe
Genomic Map