siRNA Details
| Gene Symbol: Brd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212495.1 | NP_997660.1 | 10 | 2778 | ||
| XM_006256097.2 | XP_006256159.1 | 11 | 3451 | |||
| XM_008772719.1 | XP_008770941.1 | 10 | 2148 | |||
| Gene Symbol: LOC100909544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006256037.2 | XP_006256099.1 | 10 | 3357 | ||
| XM_008772772.1 | XP_008770994.1 | 10 | 2147 | |||
| XM_008772773.1 | XP_008770995.1 | 11 | 3449 | |||

Brd2
Genomic Map