siRNA Details
| Gene Symbol: Kifc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005878.1 | NP_001005878.1 | 9 | 1956 | ||
| GenBank mRNA | BC083827.1 | - | 9 | 1956 | 7112250 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kifc1| Gene Symbol: Kifc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005878.1 | NP_001005878.1 | 9 | 1956 | ||
| GenBank mRNA | BC083827.1 | - | 9 | 1956 | 7112250 | |