siRNA Details
| Gene Symbol: Cuta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164706.1 | NP_001158178.1 | 4 | 370 | ||
| NM_001164707.1 | NP_001158179.1 | 3 | 423 | |||
| NM_212494.2 | NP_997659.2 | 4 | 423 | |||
| XM_006256120.2 | XP_006256182.1 | 5 | 454 | |||
| GenBank mRNA | BP501235.1 | - | 3 | 437 | ||
| CB586341.1 | - | 4 | 370 | |||
| DV717217.1 | - | 3 | 395 | |||
| FQ220708.1 | - | 4 | 657 | |||
| FQ223106.1 | - | 2 | 97 | |||
| FQ229047.1 | - | 2 | 97 | |||

Cuta
Genomic Map