siRNA Details
| Gene Symbol: Nudt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024243.1 | NP_001019414.1 | 4, 5 | 541 | ||
| GenBank mRNA | BC093618.1 | - | 4, 5 | 541 | 7311727 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nudt3| Gene Symbol: Nudt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024243.1 | NP_001019414.1 | 4, 5 | 541 | ||
| GenBank mRNA | BC093618.1 | - | 4, 5 | 541 | 7311727 | |