siRNA Details
| Gene Symbol: Pi16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170481.1 | NP_001163952.1 | 5 | 854 | ||
| GenBank mRNA | BC100265.1 | - | 5 | 854 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pi16| Gene Symbol: Pi16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170481.1 | NP_001163952.1 | 5 | 854 | ||
| GenBank mRNA | BC100265.1 | - | 5 | 854 | ||