siRNA Details
| Gene Symbol: Glo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207594.2 | NP_997477.1 | 1, 2 | 169 | ||
| GenBank mRNA | BC061570.1 | - | 1, 2 | 173 | 6918143 | |
| FQ210595.1 | - | 1, 2 | 147 | |||
| FQ220772.1 | - | 1, 2 | 169 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glo1| Gene Symbol: Glo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207594.2 | NP_997477.1 | 1, 2 | 169 | ||
| GenBank mRNA | BC061570.1 | - | 1, 2 | 173 | 6918143 | |
| FQ210595.1 | - | 1, 2 | 147 | |||
| FQ220772.1 | - | 1, 2 | 169 | |||