siRNA Details
| Gene Symbol: Slc37a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011944.1 | NP_001011944.1 | 6 | 762 | ||
| XM_008772807.1 | XP_008771029.1 | 4 | 495 | |||
| GenBank mRNA | BC081990.1 | - | 6 | 762 | 7129340 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc37a1| Gene Symbol: Slc37a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011944.1 | NP_001011944.1 | 6 | 762 | ||
| XM_008772807.1 | XP_008771029.1 | 4 | 495 | |||
| GenBank mRNA | BC081990.1 | - | 6 | 762 | 7129340 | |