siRNA Details
| Gene Symbol: Ube2g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106380.1 | NP_001099850.1 | 4, 5 | 323 | ||
| GenBank mRNA | BC166733.1 | - | 4, 5 | 256 | 7936238 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2g2| Gene Symbol: Ube2g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106380.1 | NP_001099850.1 | 4, 5 | 323 | ||
| GenBank mRNA | BC166733.1 | - | 4, 5 | 256 | 7936238 | |