siRNA Details
| Gene Symbol: Pcbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011945.1 | NP_001011945.1 | 4, 5 | 524 | ||
| XM_006256283.2 | XP_006256345.1 | 4, 5 | 574 | |||
| XM_006256285.2 | XP_006256347.1 | 1, 2 | 158 | |||
| XM_006256286.2 | XP_006256348.1 | 4, 5 | 574 | |||
| XM_006256287.2 | XP_006256349.1 | 4, 5 | 575 | |||
| XM_006256288.2 | XP_006256350.1 | 4, 5 | 572 | |||
| XM_006256289.2 | XP_006256351.1 | 4, 5 | 572 | |||
| XM_006256290.2 | XP_006256352.1 | 2, 3 | 1573 | |||
| XM_006256291.2 | XP_006256353.1 | 4, 5 | 573 | |||
| XM_008772874.1 | XP_008771096.1 | 2, 3 | 1594 | |||
| XM_008772875.1 | XP_008771097.1 | 2, 3 | 278 | |||
| GenBank mRNA | BC079196.1 | - | 4, 5 | 524 | 7131426 | |

Pcbp3
Genomic Map