siRNA Details
| Gene Symbol: Cisd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106385.2 | NP_001099855.1 | 3 | 361 | ||
| GenBank mRNA | BC159419.1 | - | 3 | 361 | 8370407 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cisd1| Gene Symbol: Cisd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106385.2 | NP_001099855.1 | 3 | 361 | ||
| GenBank mRNA | BC159419.1 | - | 3 | 361 | 8370407 | |