siRNA Details
| Gene Symbol: Lrrtm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106387.1 | NP_001099857.1 | 2 | 1161 | ||
| XM_008772878.1 | XP_008771100.1 | 2 | 2837 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_178678, AK035882, AK035773, BC094910, AK163753, and others.

Lrrtm3
Genomic Map