siRNA Details
| Gene Symbol: Rfx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106388.1 | NP_001099858.1 | 3, 4 | 371 | ||
| XM_006256444.2 | XP_006256506.1 | 6, 7 | 673 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rfx6| Gene Symbol: Rfx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106388.1 | NP_001099858.1 | 3, 4 | 371 | ||
| XM_006256444.2 | XP_006256506.1 | 6, 7 | 673 | |||