siRNA Details
| Gene Symbol: Man1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033656.1 | NP_001028828.1 | 7, 8 | 1646 | ||
| XM_006256497.1 | XP_006256559.1 | 7, 8 | 1518 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Man1a1| Gene Symbol: Man1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033656.1 | NP_001028828.1 | 7, 8 | 1646 | ||
| XM_006256497.1 | XP_006256559.1 | 7, 8 | 1518 | |||