siRNA Details
| Gene Symbol: Man1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033656.1 | NP_001028828.1 | 10 | 1863 | ||
| XM_006256497.1 | XP_006256559.1 | 10 | 1735 | |||
| GenBank mRNA | BC090336.1 | - | 2 | 40 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Man1a1| Gene Symbol: Man1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033656.1 | NP_001028828.1 | 10 | 1863 | ||
| XM_006256497.1 | XP_006256559.1 | 10 | 1735 | |||
| GenBank mRNA | BC090336.1 | - | 2 | 40 | ||