siRNA Details
| Gene Symbol: Ranbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191604.1 | NP_001178533.1 | 1337 | |||
| XM_006256402.2 | XP_006256464.1 | 409 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ranbp2| Gene Symbol: Ranbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191604.1 | NP_001178533.1 | 1337 | |||
| XM_006256402.2 | XP_006256464.1 | 409 | ||||