siRNA Details
| Gene Symbol: Nt5dc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106393.1 | NP_001099863.1 | 11 | 1336 | ||
| XM_008772980.1 | XP_008771202.1 | 11 | 1352 | |||
| XM_008772981.1 | XP_008771203.1 | 11 | 1349 | |||
| GenBank mRNA | BC162021.1 | - | 11 | 1300 | 9098589 | |

Nt5dc1
Genomic Map