siRNA Details
| Gene Symbol: Pald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034128.2 | NP_001029300.2 | 3 | 304 | ||
| XM_006256446.2 | XP_006256508.1 | 4 | 1079 | |||
| XM_006256447.2 | XP_006256509.1 | 4 | 515 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pald1| Gene Symbol: Pald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034128.2 | NP_001029300.2 | 3 | 304 | ||
| XM_006256446.2 | XP_006256508.1 | 4 | 1079 | |||
| XM_006256447.2 | XP_006256509.1 | 4 | 515 | |||