siRNA Details
| Gene Symbol: Ascc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007632.2 | NP_001007633.1 | 2 | 115 | ||
| XM_006256404.2 | XP_006256466.1 | 2 | 309 | |||
| XM_006256405.2 | XP_006256467.1 | 2 | 306 | |||
| XM_006256407.2 | XP_006256469.2 | 2 | 195 | |||
| XM_006256408.2 | XP_006256470.1 | 2 | 264 | |||
| XM_006256409.1 | XP_006256471.1 | 1 | 50 | |||
| XM_006256410.2 | XP_006256472.1 | 2 | 261 | |||
| XM_008772881.1 | XP_008771103.1 | 1 | 47 | |||
| GenBank mRNA | CK357599.1 | - | 2 | 115 | ||

Ascc1
Genomic Map