siRNA Details
| Gene Symbol: Cep57l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017448.1 | NP_001017448.1 | 9 | 1031 | ||
| XM_006256562.2 | XP_006256624.1 | 9 | 1312 | |||
| XM_006256563.2 | XP_006256625.1 | 9 | 1267 | |||
| XM_006256564.2 | XP_006256626.1 | 9 | 1067 | |||
| XM_006256565.1 | XP_006256627.1 | 10 | 1090 | |||
| XM_006256566.2 | XP_006256628.1 | 9 | 1724 | |||
| XM_006256567.2 | XP_006256629.1 | 10 | 1059 | |||
| XM_006256568.2 | XP_006256630.1 | 8 | 942 | |||
| XM_008772982.1 | XP_008771204.1 | 9 | 1982 | |||
| XM_008772983.1 | XP_008771205.1 | 9 | 1055 | |||
| GenBank mRNA | BC079256.1 | - | 9 | 1031 | 7133287 | |

Cep57l1
Genomic Map