siRNA Details
| Gene Symbol: Pou5f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009178.2 | NP_001009178.1 | 5 | 937 | ||
| GenBank mRNA | BC158566.1 | - | 5 | 937 | 9092936 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pou5f1| Gene Symbol: Pou5f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009178.2 | NP_001009178.1 | 5 | 937 | ||
| GenBank mRNA | BC158566.1 | - | 5 | 937 | 9092936 | |