siRNA Details
| Gene Symbol: Fam13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772939.1 | XP_008771161.1 | 3 | 7178 | ||
| XM_008772940.1 | XP_008771162.1 | 3 | 7178 | |||
| GenBank mRNA | FQ212194.1 | - | 942 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam13c| Gene Symbol: Fam13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772939.1 | XP_008771161.1 | 3 | 7178 | ||
| XM_008772940.1 | XP_008771162.1 | 3 | 7178 | |||
| GenBank mRNA | FQ212194.1 | - | 942 | |||