siRNA Details
| Gene Symbol: Btf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008309.1 | NP_001008310.1 | 3, 4 | 299 | ||
| XM_006231835.2 | XP_006231897.1 | 3, 4 | 569 | |||
| GenBank mRNA | BC085343.1 | - | 3, 4 | 299 | 7303981 | |
| FQ220452.1 | - | 3, 4 | 303 | |||
| FQ229173.1 | - | 3, 4 | 305 | |||

Btf3
Genomic Map