siRNA Details
| Gene Symbol: Sgtb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181629.3 | NP_853660.1 | 7 | 645 | ||
| XM_006231882.2 | XP_006231944.1 | 7 | 811 | |||
| XM_006231883.2 | XP_006231945.1 | 7 | 786 | |||
| GenBank mRNA | AF368280.1 | - | 7 | 646 | ||
| FQ211527.1 | - | 7 | 689 | |||
| FQ212077.1 | - | 7 | 813 | |||
| FQ212215.1 | - | 7 | 788 | |||

Sgtb
Genomic Map