siRNA Details
| Gene Symbol: Ppwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106406.1 | NP_001099876.1 | 5 | 1094 | ||
| XM_006231887.2 | XP_006231949.1 | 5 | 871 | |||
| XM_008760666.1 | XP_008758888.1 | 7 | 866 | |||
| GenBank mRNA | BC078983.1 | - | 5 | 1299 | ||

Ppwd1
Genomic Map