siRNA Details
| Gene Symbol: Cenpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106407.1 | NP_001099877.1 | 6 | 749 | ||
| XM_006231888.1 | XP_006231950.1 | 6 | 749 | |||
| XM_006231889.2 | XP_006231951.1 | 6 | 470 | |||
| XM_006231890.2 | XP_006231952.1 | 6 | 378 | |||
| XM_008760667.1 | XP_008758889.1 | 6 | 749 | |||

Cenpk
Genomic Map