siRNA Details
| Gene Symbol: Card6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106413.1 | NP_001099883.1 | 3 | 1486 | ||
| XM_008760760.1 | XP_008758982.1 | 3 | 1613 | |||
| XM_008760761.1 | XP_008758983.1 | 4 | 1536 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Card6| Gene Symbol: Card6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106413.1 | NP_001099883.1 | 3 | 1486 | ||
| XM_008760760.1 | XP_008758982.1 | 3 | 1613 | |||
| XM_008760761.1 | XP_008758983.1 | 4 | 1536 | |||