siRNA Details
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013909.1 | NP_001013931.1 | 17 | 1777 | ||
GenBank mRNA | BC089903.1 | - | 17 | 1777 | 7384104 | |
FQ214211.1 | - | 2 | 108 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013909.1 | NP_001013931.1 | 17 | 1777 | ||
GenBank mRNA | BC089903.1 | - | 17 | 1777 | 7384104 | |
FQ214211.1 | - | 2 | 108 |