siRNA Details
| Gene Symbol: Capsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106417.1 | NP_001099887.1 | 4 | 542 | ||
| XM_008760763.1 | XP_008758985.1 | 4 | 518 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Capsl| Gene Symbol: Capsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106417.1 | NP_001099887.1 | 4 | 542 | ||
| XM_008760763.1 | XP_008758985.1 | 4 | 518 | |||