siRNA Details
Gene Symbol: Rxfp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008310.1 | NP_001008311.1 | 1 | 865 | ||
GenBank mRNA | AY633763.1 | - | 1 | 844 | ||
AY633764.1 | - | 1 | 865 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
